The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291222.1 | NP_001278151.1 | 1 - 2 | 359 | 80 |
NM_010626.3 | NP_034756.2 | 1 - 2 | 359 | 80 | |
GenBank mRNA | AB001435.1 | - | 1 - 2 | 64 | 80 |
BC141045.1 | - | 1 - 2 | 360 | 80 | |
BC145222.1 | - | 1 - 2 | 360 | 80 |