The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007926.2 | NP_031952.2 | 3 - 4 | 333 | 146 |
GenBank mRNA | AK087932.1 | - | 3 - 4 | 288 | 146 |
AK133859.1 | - | 3 - 4 | 286 | 146 | |
BC002054.1 | - | 3 - 4 | 288 | 146 | |
BY256548.1 | - | 3 - 4 | 338 | 146 | |
U10118.1 | - | 3 - 4 | 284 | 146 |