Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sash3
Entrez Gene ID
74131
Gene Name
SAM and SH3 domain containing 3
Gene Aliases
1200013B08Rik, AW413946, SLY1
Chromosome Location
Chr.X: 48146451 - 48161565 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028773.3 NP_083049.1 2 - 3 286 93
GenBank mRNA AJ306422.1 - 2 - 3 152 93
AK004734.2 - 2 - 3 287 93
AK156202.1 - 2 - 3 351 93
AK171017.1 - 2 - 3 284 93
BC064734.1 - 2 - 3 271 93

View Full Product Details