Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aifm1
Entrez Gene ID
26926
Gene Name
apoptosis-inducing factor, mitochondrion-associated 1
Gene Aliases
AIF, AIFsh2, Hq, Pdcd8
Chromosome Location
Chr.X: 48474944 - 48513563 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012019.3 NP_036149.1 2 - 3 464 115
GenBank mRNA AF100927.1 - 2 - 3 290 115
AK133764.1 - 2 - 3 305 115
BC003292.1 - 2 - 3 327 115
BC024882.1 - 2 - 3 948 115
DQ016499.1 - 2 - 3 327 115

View Full Product Details