The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003909.4 | NP_001003909.2 | 19 - 20 | 3126 | 75 |
NM_001289527.1 | NP_001276456.1 | 19 - 20 | 3144 | 75 | |
NM_001289528.1 | NP_001276457.1 | 18 - 19 | 3008 | 75 | |
NM_001289529.1 | NP_001276458.1 | 19 - 20 | 2753 | 75 | |
GenBank mRNA | AK129343.1 | - | 19 - 20 | 2927 | 75 |
BC079620.1 | - | 8 - 9 | 816 | 75 | |
BC089031.1 | - | 19 - 20 | 2732 | 75 | |
BC145510.1 | - | 18 - 19 | 2759 | 75 | |
BC145511.1 | - | 19 - 20 | 2895 | 75 | |
BC150792.1 | - | 19 - 20 | 2895 | 75 |