Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankib1
Entrez Gene ID
70797
Gene Name
ankyrin repeat and IBR domain containing 1
Gene Aliases
2310061P20Rik, 4631416I11Rik, AW494740, C80642, mKIAA1386
Chromosome Location
Chr.5: 3689999 - 3803124 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003909.4 NP_001003909.2 3 - 4 1092 85
NM_001289527.1 NP_001276456.1 3 - 4 1092 85
NM_001289528.1 NP_001276457.1 3 - 4 1092 85
NM_001289529.1 NP_001276458.1 2 - 3 564 85
GenBank mRNA AK129343.1 - 3 - 4 893 85
BC020475.1 - 3 - 4 626 85
BC089031.1 - 2 - 3 543 85
BC145510.1 - 3 - 4 843 85
BC145511.1 - 3 - 4 843 85
BC150792.1 - 3 - 4 843 85

View Full Product Details