The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033285.1 | NP_001028457.1 | 19 - 20 | 2754 | 96 |
GenBank mRNA | AF492452.1 | - | 5 - 6 | 528 | 96 |
AK044466.1 | - | 1 - 2 | 160 | 96 | |
AK047837.1 | - | 3 - 4 | 2516 | 96 | |
BC158017.1 | - | 19 - 20 | 2754 | 96 | |
BC158095.1 | - | 19 - 20 | 2754 | 96 |