Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdc42bpa
Entrez Gene ID
226751
Gene Name
CDC42 binding protein kinase alpha
Gene Aliases
A930014J19Rik, DMPK-like
Chromosome Location
Chr.1: 179960093 - 180165606 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033285.1 NP_001028457.1 36 - 37 5087 86
GenBank mRNA AK129148.1 - 7 - 8 902 86
BC096679.1 - 6 - 7 679 86
BC158017.1 - 36 - 37 5087 86
BC158095.1 - 35 - 36 5048 86

View Full Product Details