Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dlgap5
Entrez Gene ID
218977
Gene Name
discs, large (Drosophila) homolog-associated protein 5
Gene Aliases
C77459, C86398, Dap-5, Dlg7, Hurp, mKIAA0008
Chromosome Location
Chr.14: 47387779 - 47418407 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144553.2 NP_653136.2 13 - 14 1890 83
GenBank mRNA AB076696.1 - 13 - 14 1890 83
AK036384.1 - 13 - 14 1692 83
AK048470.1 - 13 - 14 1660 83
AK085882.1 - 13 - 14 1671 83
AK135820.1 - 13 - 14 1697 83
AK147955.1 - 13 - 14 1660 83
BC058087.1 - 13 - 14 1624 83

View Full Product Details