Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gmfb
Entrez Gene ID
63985
Gene Name
glia maturation factor, beta
Gene Aliases
3110001H22Rik, 3110001O16Rik, AI851627, C79176, D14Ertd630e
Chromosome Location
Chr.14: 46808149 - 46822263 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB001732.1 - 2 - 3 164 114
AF297220.1 - 2 - 3 210 114

View Full Product Details