The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173863.2 | NP_776288.2 | 10 - 11 | 1107 | 81 |
GenBank mRNA | AK144840.1 | - | 10 - 11 | 1109 | 81 |
BC011210.1 | - | 10 - 11 | 946 | 81 |