The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253755.1 | NP_001240684.1 | 5 - 6 | 717 | 90 |
NM_172605.3 | NP_766193.3 | 5 - 6 | 717 | 90 | |
GenBank mRNA | AK032892.1 | - | 5 - 6 | 662 | 90 |
AK033390.1 | - | 5 - 6 | 662 | 90 | |
BC005670.1 | - | 5 - 6 | 618 | 90 | |
BC066144.1 | - | 5 - 6 | 665 | 90 |