Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd42
Entrez Gene ID
73845
Gene Name
ankyrin repeat domain 42
Gene Aliases
4931426M20, 4933417L02Rik, Ikbn
Chromosome Location
Chr.7: 92584175 - 92637143 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028665.4 NP_082941.2 9 - 10 2141 100
GenBank mRNA AK016847.1 - 4 - 5 422 100
AK029511.1 - 9 - 10 2143 100
AK029888.1 - 9 - 10 2102 100
AK133339.1 - - 2102 100
BC030885.1 - 9 - 10 1770 100
BC100433.1 - 9 - 10 2177 100
CV563396.1 - 3 - 4 100

View Full Product Details