The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177591.4 | NP_808259.2 | 2 - 3 | 222 | 94 |
NM_177915.4 | NP_808583.1 | 2 - 3 | 222 | 94 | |
NM_183335.2 | NP_899178.2 | 1 - 2 | 86 | 94 | |
GenBank mRNA | AK030452.1 | - | 2 - 3 | 194 | 94 |
AK129126.1 | - | 2 - 3 | 204 | 94 | |
AY227771.1 | - | 2 - 3 | 216 | 94 | |
AY227772.1 | - | 2 - 3 | 216 | 94 | |
AY227773.1 | - | 1 - 2 | 86 | 94 | |
BC089474.1 | - | 2 - 3 | 203 | 94 | |
BC100420.1 | - | 2 - 3 | 222 | 94 |