Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prdm12
Entrez Gene ID
381359
Gene Name
PR domain containing 12
Gene Aliases
Gm998
Chromosome Location
Chr.2: 31633517 - 31655795 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123362.1 NP_001116834.1 2 - 3 481 95

View Full Product Details