Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lamc3
Entrez Gene ID
23928
Gene Name
laminin gamma 3
Gene Aliases
AI562206, AW240805
Chromosome Location
Chr.2: 31887281 - 31946535 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011836.3 NP_035966.2 2 - 3 768 91
GenBank mRNA AF079520.1 - 2 - 3 758 91
AF083372.1 - 2 - 3 705 91
BC096366.1 - 2 - 3 768 91

View Full Product Details