The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009225.2 | NP_033251.1 | 6 - 7 | 813 | 103 |
GenBank mRNA | AK077537.1 | - | 6 - 7 | 767 | 103 |
AK146319.1 | - | 6 - 7 | 856 | 103 |