The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289747.1 | NP_001276676.1 | 8 - 9 | 1218 | 64 |
NM_001289748.1 | NP_001276677.1 | 7 - 8 | 1055 | 64 | |
NM_001289749.1 | NP_001276678.1 | 5 - 6 | 712 | 64 | |
NM_177726.4 | NP_808394.1 | 7 - 8 | 1074 | 64 | |
GenBank mRNA | AK037229.1 | - | 7 - 8 | 1064 | 64 |
AY159126.1 | - | 7 - 8 | 1064 | 64 | |
AY177606.1 | - | 5 - 6 | 702 | 64 | |
AY177607.1 | - | 7 - 8 | 1055 | 64 | |
BC112420.1 | - | 7 - 8 | 1051 | 64 | |
BC113796.1 | - | 8 - 9 | 1195 | 64 |