Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psmd14
Entrez Gene ID
59029
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 14
Gene Aliases
2610312C03Rik, 3200001M20Rik, AA986732, Pad1, Poh1, rpm11
Chromosome Location
Chr.2: 61711694 - 61800376 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021526.2 NP_067501.2 8 - 9 883 141
GenBank mRNA AK012013.1 - 8 - 9 814 141
AK031863.1 - 8 - 9 820 141
AK078134.1 - 8 - 9 835 141
AK150280.1 - 8 - 9 819 141
AK151104.1 - 8 - 9 866 141
AK151733.1 - 8 - 9 866 141
BC003742.1 - 8 - 9 788 141
Y13071.1 - 8 - 9 804 141

View Full Product Details