The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172152.2 | NP_742164.1 | 14 - 15 | 1513 | 71 |
GenBank mRNA | AK044239.1 | - | 14 - 15 | 2145 | 71 |
AY156046.1 | - | 14 - 15 | 1513 | 71 | |
BC137945.1 | - | 14 - 15 | 1484 | 71 | |
BC171939.1 | - | 14 - 15 | 1484 | 71 |