The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172152.2 | NP_742164.1 | 7 - 8 | 626 | 163 |
GenBank mRNA | AK044239.1 | - | 7 - 8 | 1315 | 163 |
AY156046.1 | - | 7 - 8 | 626 | 163 | |
BC137945.1 | - | 7 - 8 | 654 | 163 | |
BC171939.1 | - | 7 - 8 | 654 | 163 |