Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oca2
Entrez Gene ID
18431
Gene Name
oculocutaneous albinism II
Gene Aliases
D7H15S12, D7Icr28RN, D7Nic1, p
Chromosome Location
Chr.7: 56239593 - 56536517 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021879.2 NP_068679.1 3 - 4 447 86
GenBank mRNA AK051897.1 - 3 - 4 442 86
BC119220.1 - 3 - 4 417 86
BC120549.1 - 3 - 4 417 86
M97900.1 - 3 - 4 447 86

View Full Product Details