The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177610.2 | NP_808278.2 | 27 - 28 | 3754 | 70 |
GenBank mRNA | BC172138.1 | - | 28 - 29 | 4130 | 70 |