Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Surf2
Entrez Gene ID
20931
Gene Name
surfeit gene 2
Gene Aliases
Surf-2
Chromosome Location
Chr.2: 26916372 - 26920183 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013678.2 NP_038706.1 3 - 4 351 68
GenBank mRNA AK157125.1 - 3 - 4 365 68
AK165559.1 - 3 - 4 358 68
AK166181.1 - 3 - 4 380 68
AK172018.1 - 3 - 4 354 68
BC096401.1 - 3 - 4 351 68

View Full Product Details