The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138942.3 | NP_620392.2 | 11 - 12 | 1743 | 68 |
GenBank mRNA | AK132245.1 | - | 11 - 12 | 1744 | 68 |
BC141022.1 | - | 11 - 12 | 1770 | 68 | |
BC171949.1 | - | 11 - 12 | 1770 | 68 |