The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008498.2 | NP_001008498.2 | 3 - 4 | 553 | 83 |
GenBank mRNA | AK014304.1 | - | 3 - 4 | 552 | 83 |
BC085113.1 | - | 3 - 4 | 509 | 83 |