The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026615.4 | NP_080891.1 | 4 - 5 | 250 | 79 |
GenBank mRNA | AK012124.1 | - | 4 - 5 | 256 | 79 |
AK019352.1 | - | 4 - 5 | 256 | 79 | |
AK049227.1 | - | 4 - 5 | 251 | 79 | |
AK049849.1 | - | 4 - 5 | 251 | 79 | |
BC026994.1 | - | 4 - 5 | 242 | 79 |