Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pacs2
Entrez Gene ID
217893
Gene Name
phosphofurin acidic cluster sorting protein 2
Gene Aliases
6720425G15Rik, AW208651, AW489976, E230011J22, Pacs1l, mKIAA0602
Chromosome Location
Chr.12: 113014507 - 113074401 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081170.1 NP_001074639.1 5 - 6 636 61
NM_001291444.1 NP_001278373.1 5 - 6 636 61
NM_001291445.1 NP_001278374.1 4 - 5 546 61
GenBank mRNA AK036581.1 - 4 - 5 522 61
AK054000.1 - 5 - 6 611 61
AK122326.1 - 3 - 4 325 61

View Full Product Details