Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snapc4
Entrez Gene ID
227644
Gene Name
small nuclear RNA activating complex, polypeptide 4
Gene Aliases
5730436L13Rik
Chromosome Location
Chr.2: 26362765 - 26381072 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290419.1 NP_001277348.1 10 - 11 1039 115
NM_172339.4 NP_758842.1 9 - 10 920 115
GenBank mRNA AK077522.1 - 9 - 10 887 115
AK136996.1 - 9 - 10 897 115
BC018169.1 - 9 - 10 886 115
BC057031.1 - 10 - 11 1005 115

View Full Product Details