The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285458.1 | NP_001272387.1 | 14 - 15 | 3933 | 90 |
NM_001285459.1 | NP_001272388.1 | 14 - 15 | 3557 | 90 | |
NM_001287817.1 | NP_001274746.1 | 2 - 3 | 1437 | 90 | |
NM_001287818.1 | NP_001274747.1 | 3 - 4 | 177 | 90 | |
NM_010230.2 | NP_034360.2 | 17 - 18 | 4335 | 90 | |
GenBank mRNA | AK076603.1 | - | 2 - 3 | 1423 | 90 |
BC138036.1 | - | 13 - 14 | 3839 | 90 | |
BC171945.1 | - | 13 - 14 | 3833 | 90 | |
X53599.1 | - | 17 - 18 | 4295 | 90 | |
X62379.1 | - | 14 - 15 | 3563 | 90 |