Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wars
Entrez Gene ID
22375
Gene Name
tryptophanyl-tRNA synthetase
Gene Aliases
TrpRS, WRS
Chromosome Location
Chr.12: 108860030 - 108894174 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011710.3 NP_035840.3 10 - 11 1537 56

View Full Product Details