Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phospho2
Entrez Gene ID
73373
Gene Name
phosphatase, orphan 2
Gene Aliases
1700048E23Rik, AI661517, AU021728, AV006103, Phos2
Chromosome Location
Chr.2: 69789628 - 69797223 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028521.2 NP_082797.1 2 - 3 251 101
GenBank mRNA AK006724.1 - 2 - 3 252 101
AK010147.1 - - 277 101
AK159551.1 - 2 - 3 299 101

View Full Product Details