The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031621.2 | NP_001026792.2 | 26 - 27 | 4502 | 62 |
GenBank mRNA | AB112584.1 | - | 26 - 27 | 4502 | 62 |