The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034874.3 | NP_001030046.1 | 1 - 2 | 820 | 61 |
GenBank mRNA | AK147504.1 | - | 1 - 2 | 821 | 61 |