The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175681.3 | NP_783612.2 | 12 - 13 | 1497 | 56 |
GenBank mRNA | AY605231.1 | - | 12 - 13 | 1216 | 56 |
BC044746.2 | - | 12 - 13 | 1497 | 56 |