Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Glp2r
Entrez Gene ID
93896
Gene Name
glucagon-like peptide 2 receptor
Gene Aliases
9530092J08Rik, GLP-2
Chromosome Location
Chr.11: 67663299 - 67771153 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175681.3 NP_783612.2 4 - 5 665 65
GenBank mRNA AK035702.1 - - 214 65
AY255567.1 - 3 - 4 279 65
AY605231.1 - 4 - 5 384 65
BC044746.2 - 4 - 5 665 65

View Full Product Details