Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Snhg7
- Entrez Gene ID
-
72091
- Gene Name
- small nucleolar RNA host gene 7
- Gene Aliases
-
2610002F03Rik, AI427062
- Chromosome Location
-
Chr.2: 26637176 - 26640244 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024068.2 |
|
3 - 4 |
291 |
82 |
GenBank mRNA |
AK011283.1 |
- |
3 - 4 |
292 |
82 |
AK028166.1 |
- |
3 - 4 |
292 |
82 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY710585.1 |
- |
3 - 4 |
288 |
|
Chr. 2 - 26637229 - 26640241 [-] on Build GRCm38 |
82 |
GenBankmRNA |
BY710095.1 |
- |
3 - 4 |
286 |
|
Chr. 2 - 26637299 - 26640239 [-] on Build GRCm38 |
82 |
GenBankmRNA |
CA466784.1 |
- |
3 - 4 |
328 |
|
Chr. 2 - 26637217 - 26640244 [-] on Build GRCm38 |
82 |
GenBankmRNA |
AA041850.1 |
- |
2 - 3 |
118 |
|
Chr. 2 - 26637257 - 26640113 [-] on Build GRCm38 |
82 |
GenBankmRNA |
AA982539.1 |
- |
3 - 4 |
263 |
|
Chr. 2 - 26637475 - 26640210 [-] on Build GRCm38 |
82 |
GenBankmRNA |
AV572272.1 |
- |
3 - 4 |
276 |
|
Chr. 2 - 26637440 - 26640229 [-] on Build GRCm38 |
82 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
small nucleolar RNA, H/ACA box 43 |
Snora43 |
ACA43 |
|
Sense |
Chr. - 26637847 - 26637985 on Build GRCm38 |
Downstream |
small nucleolar RNA, H/ACA box 17 |
Snora17 |
ACA17 |
1.1kb
|
Sense |
Chr. - 26639190 - 26639321 on Build GRCm38 |
View Full Product Details