Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lcn3
Entrez Gene ID
16820
Gene Name
lipocalin 3
Gene Aliases
Vnsp1
Chromosome Location
Chr.2: 25765569 - 25768099 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010694.1 NP_034824.1 2 - 3 222 87
GenBank mRNA BC116816.1 - 2 - 3 243 87
BC116818.1 - 2 - 3 243 87
BC144882.1 - 2 - 3 243 87
D38580.1 - 2 - 3 222 87

View Full Product Details