Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- C530044C16Rik
- Entrez Gene ID
-
319981
- Gene Name
- RIKEN cDNA C530044C16 gene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 50776115 - 50814894 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_045984.1 |
|
2 - 3 |
166 |
89 |
GenBank mRNA |
AK043004.1 |
- |
2 - 3 |
167 |
89 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BB640265.1 |
- |
2 - 3 |
167 |
|
Chr. 6 - 50776115 - 50814334 [+] on Build GRCm38 |
89 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
neuropeptide VF precursor |
Npvf |
Rfrp |
-121.9kb
|
Anti-sense |
Chr. - 50650871 - 50654393 on Build GRCm38 |
Downstream |
predicted gene, 31579 |
Gm31579 |
|
109.5kb
|
Sense |
Chr. - 50906401 - 50910734 on Build GRCm38 |
View Full Product Details