The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013802.3 | NP_001013824.2 | 3 - 4 | 517 | 57 |
GenBank mRNA | AK013491.1 | - | 3 - 4 | 514 | 57 |
AK134694.1 | - | 3 - 4 | 519 | 57 |