Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prrx2
Entrez Gene ID
20204
Gene Name
paired related homeobox 2
Gene Aliases
Prx2, S8
Chromosome Location
Chr.2: 30845064 - 30881251 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009116.2 NP_033142.2 1 - 2 319 79
GenBank mRNA BC137872.1 - 1 - 2 346 79
BC137874.1 - 1 - 2 346 79
CJ179520.1 - 1 - 2 320 79

View Full Product Details