Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kcnab3
Entrez Gene ID
16499
Gene Name
potassium voltage-gated channel, shaker-related subfamily, beta member 3
Gene Aliases
C330022D06Rik, Kcnab4, mKv(beta)4
Chromosome Location
Chr.11: 69326258 - 69333041 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010599.4 NP_034729.3 12 - 13 1474 108
GenBank mRNA AK029750.1 - 11 - 12 1064 108
AK031806.1 - 12 - 13 1455 108
AK044102.1 - 12 - 13 1429 108
AK046194.1 - 12 - 13 1464 108
AK083134.1 - 12 - 13 1476 108
BC017518.1 - 12 - 13 1400 108

View Full Product Details