Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klc1
Entrez Gene ID
16593
Gene Name
kinesin light chain 1
Gene Aliases
AI874768, Kns2
Chromosome Location
Chr.12: 111758849 - 111807844 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025358.2 NP_001020529.2 4 - 5 751 97
NM_001025359.2 NP_001020530.2 4 - 5 751 97
NM_001025360.2 NP_001020531.2 4 - 5 770 97
NM_001025361.2 NP_001020532.2 4 - 5 751 97
NM_001025362.2 NP_001020533.2 4 - 5 751 97
NM_001025363.2 NP_001020534.2 4 - 5 751 97
NM_001081959.1 NP_001075428.1 4 - 5 770 97
NM_008450.2 NP_032476.2 4 - 5 770 97
GenBank mRNA AK031309.1 - 4 - 5 742 97
AY753300.1 - 3 - 4 575 97
BC055744.1 - 4 - 5 606 97

View Full Product Details