The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025358.2 | NP_001020529.2 | 2 - 3 | 435 | 74 |
NM_001025359.2 | NP_001020530.2 | 2 - 3 | 435 | 74 | |
NM_001025360.2 | NP_001020531.2 | 2 - 3 | 454 | 74 | |
NM_001025361.2 | NP_001020532.2 | 2 - 3 | 435 | 74 | |
NM_001025362.2 | NP_001020533.2 | 2 - 3 | 435 | 74 | |
NM_001025363.2 | NP_001020534.2 | 2 - 3 | 435 | 74 | |
NM_001081959.1 | NP_001075428.1 | 2 - 3 | 454 | 74 | |
NM_008450.2 | NP_032476.2 | 2 - 3 | 454 | 74 | |
GenBank mRNA | AF055665.1 | - | 2 - 3 | 353 | 74 |
AK031309.1 | - | 2 - 3 | 426 | 74 | |
AK197521.1 | - | 1 - 2 | 43 | 74 | |
AU035615.1 | - | 2 - 3 | 454 | 74 | |
AY753300.1 | - | 1 - 2 | 259 | 74 | |
BC055744.1 | - | 2 - 3 | 290 | 74 | |
CF615940.1 | - | 2 - 3 | 365 | 74 |