Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dclre1c
Entrez Gene ID
227525
Gene Name
DNA cross-link repair 1C, PSO2 homolog (S. cerevisiae)
Gene Aliases
9930121L06Rik, AI661365, Art, Snm1l
Chromosome Location
Chr.2: 3424062 - 3474330 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110214.1 NP_001103684.1 9 - 10 869 102
NM_146114.3 NP_666226.2 9 - 10 869 102
NM_175683.4 NP_783614.1 9 - 10 869 102
GenBank mRNA AF387731.1 - 9 - 10 851 102
AK037126.1 - 9 - 10 836 102
AK052369.1 - 9 - 10 825 102
AK052490.1 - 9 - 10 808 102
AK088810.1 - 7 - 8 651 102
AK166324.1 - 9 - 10 812 102
BC108935.2 - 9 - 10 822 102

View Full Product Details