The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040682.1 | NP_001035772.1 | 6 - 7 | 729 | 116 |
NM_053155.2 | NP_444385.2 | 6 - 7 | 729 | 116 | |
NR_104435.1 | 6 - 7 | 729 | 116 | ||
NR_104436.1 | 6 - 7 | 729 | 116 | ||
NR_104437.1 | 6 - 7 | 729 | 116 | ||
NR_104438.1 | 6 - 7 | 729 | 116 | ||
NR_104439.1 | 6 - 7 | 729 | 116 | ||
GenBank mRNA | AB047978.1 | - | 6 - 7 | 722 | 116 |
AB059643.1 | - | 6 - 7 | 722 | 116 | |
AB059644.1 | - | 6 - 7 | 722 | 116 | |
AB059645.1 | - | 6 - 7 | 722 | 116 | |
AB059646.1 | - | 6 - 7 | 722 | 116 | |
AB059647.1 | - | 6 - 7 | 722 | 116 | |
AB059648.1 | - | 6 - 7 | 722 | 116 | |
AK077023.1 | - | 3 - 4 | 311 | 116 | |
AK147777.1 | - | 6 - 7 | 626 | 116 | |
BC019176.1 | - | 6 - 7 | 660 | 116 | |
BC096470.1 | - | 6 - 7 | 618 | 116 | |
BC141280.1 | - | 6 - 7 | 662 | 116 |