Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nol7
Entrez Gene ID
70078
Gene Name
nucleolar protein 7
Gene Aliases
2210008F15Rik, 5730556I21Rik, NOP27, RARG-1
Chromosome Location
Chr.13: 43398376 - 43402858 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023554.2 NP_076043.2 2 - 3 342 55
GenBank mRNA AK008670.1 - 2 - 3 327 55
AK019972.1 - 2 - 3 327 55
AK032963.1 - 2 - 3 328 55
AK207964.1 - 2 - 3 97 55
BC090637.1 - 2 - 3 315 55
BC130012.1 - 2 - 3 332 55

View Full Product Details