Assay Design
The assay probe spans an exon junction.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BB584777.2 |
- |
1 - 2 |
83 |
|
Chr. 6 - 50800368 - 50803025 [-] on Build GRCm38 |
99 |
GenBankmRNA |
AK033863.1 |
- |
|
89 |
|
Chr. - - [-] on Build |
99 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
neuropeptide VF precursor |
Npvf |
Rfrp |
-146.0kb
|
Sense |
Chr. - 50650871 - 50654393 on Build GRCm38 |
Downstream |
predicted gene, 31579 |
Gm31579 |
|
103.4kb
|
Anti-sense |
Chr. - 50906401 - 50910734 on Build GRCm38 |
View Full Product Details