The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198977.1 | NP_001185906.1 | 11 - 12 | 1777 | 61 |
NM_011518.2 | NP_035648.2 | 11 - 12 | 1781 | 61 | |
GenBank mRNA | AK049207.1 | - | 11 - 12 | 1806 | 61 |
AK136363.1 | - | 11 - 12 | 1780 | 61 | |
AK143564.1 | - | 11 - 12 | 1810 | 61 | |
BC065121.1 | - | 11 - 12 | 1713 | 61 | |
U25685.1 | - | 10 - 11 | 1565 | 61 | |
U36776.1 | - | 11 - 12 | 1784 | 61 | |
Z49877.1 | - | 11 - 12 | 2039 | 61 |