Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cks2
- Entrez Gene ID
-
66197
- Gene Name
- CDC28 protein kinase regulatory subunit 2
- Gene Aliases
-
1110038L14Rik, CKSHS2
- Chromosome Location
-
Chr.13: 51645232 - 51650662 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK077690.1 |
- |
1 - 2 |
53 |
87 |
View Full Product Details